Stampa…

1-(2,6-Difluorobenzoyl)piperidin-4-one 95+%

Fornitore: Apollo Scientific

PC446094-1G PC446094-500MG PC446094-5G
APOSPC446094-1GEA 265 EUR
APOSPC446094-1G APOSPC446094-500MG APOSPC446094-5G
1-(2,6-Difluorobenzoyl)piperidin-4-one 95+%
1-(2,6-Difluorobenzoyl)piperidin-4-one
Formula: C₁₂H₁₁F₂NO₂
Peso molecolare: 239,218 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09942573
CAS Number: 948853-53-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR