Stampa…

2-Bromo-5-(trifluoromethyl)thiazole 95%

Fornitore: Apollo Scientific
Pericolo

PC421056-1G PC421056-250MG
APOSPC421056-1GEA 424 EUR
APOSPC421056-1G APOSPC421056-250MG
2-Bromo-5-(trifluoromethyl)thiazole 95%
2-Bromo-5-(trifluoromethyl)thiazole
Formula: C₄HBrF₃NS
Peso molecolare: 232,022 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD14702723
CAS Number: 1209458-80-9
UN: 2810
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR