Stampa…

1-Amino-3,3-difluoro-cyclobutanecarboxylic acid 97%

Fornitore: Apollo Scientific

Synonyms: 1-Amino-3,3-difluorocyclobutanecarboxylic acid

PC420003-1G PC420003-250MG
APOSPC420003-1GEA 691 EUR
APOSPC420003-1G APOSPC420003-250MG
1-Amino-3,3-difluoro-cyclobutanecarboxylic acid 97%
1-Amino-3,3-difluoro-cyclobutanecarboxylic acid
Formula: C₅H₇F₂NO₂
Peso molecolare: 151,111 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD17480490
CAS Number: 1225532-86-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR