Stampa…

3-Fluoro-5-morpholinophenylboronic acid 98%

Fornitore: Apollo Scientific

PC412143-250MG PC412143-1G
APOSPC412143-250MGEA 82.9 EUR
APOSPC412143-250MG APOSPC412143-1G
3-Fluoro-5-morpholinophenylboronic acid 98%
3-Fluoro-5-morpholinophenylboronic acid
Formula: C₁₀H₁₃BFNO₃
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12546541
CAS Number: 1217500-95-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR