3-(Difluoromethoxy)aniline hydrochloride
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₇H₇F₂NO·ClH Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD25371275 CAS Number: 1431965-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...