Stampa…

2-Amino-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid hydrochloride

Fornitore: Apollo Scientific
Attenzione

PC405702-1G PC405702-250MG
APOSPC405702-1GEA 2790 EUR
APOSPC405702-1G APOSPC405702-250MG
2-Amino-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid hydrochloride
2-Amino-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid hydrochloride
Formula: C₇H₁₀FNO₂·ClH
Peso molecolare: 195,619 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD29060099
CAS Number: 1934367-59-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR