3-Bromo-2-chloro-6-fluorobenzaldehyde
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₇H₃BrClFO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD11110260 CAS Number: 1114809-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...