Stampa…

2,4-Dichloro-3-fluorobenzenesulphonyl chloride

Fornitore: Apollo Scientific
Pericolo

PC37869-1G PC37869-250MG PC37869-5G
APOSPC37869-1GEA 575 EUR
APOSPC37869-1G APOSPC37869-250MG APOSPC37869-5G
2,4-Dichloro-3-fluorobenzenesulphonyl chloride
2,4-Dichloro-3-fluorobenzenesulphonyl chloride
Formula: C₆H₂Cl₃FO₂S
Peso molecolare: 263,501 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD20233369
CAS Number: 1349718-19-9
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR