Ethyl trifluoroacetyldibromoacetate 96%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₆H₅Br₂F₃O₃ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD07784253 CAS Number: 382-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...