3-(3-Fluorobenzoyl)-1H-pyrrole
Fornitore: Apollo Scientific
|
Pericolo
|
Synonyms:
(3-Fluorophenyl)-(1H-pyrrol-3-yl)methanone
PC3157-1G
PC3157-500MG
APOSPC3157-1GEA
695
EUR
APOSPC3157-1G
APOSPC3157-500MG
3-(3-Fluorobenzoyl)-1H-pyrrole
3-(3-Fluorobenzoyl)-1H-pyrrole
Formula:
C₁₁H₈FNO Peso molecolare: 189,186 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD08056193 CAS Number: 893612-64-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...