Stampa…

3-Bromo-2,5,6-trifluorobenzoyl chloride

Fornitore: Apollo Scientific

PC303511-1G PC303511-5G
APOSPC303511-1GEA 232 EUR
APOSPC303511-1G APOSPC303511-5G
3-Bromo-2,5,6-trifluorobenzoyl chloride
3-Bromo-2,5,6-trifluorobenzoyl chloride
Formula: C₇HBrClF₃O
Peso molecolare: 273,434 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD32640103
CAS Number: 1518390-38-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR