Stampa…

4,6-Dichloro-2-fluoro-3-methoxybenzylamine

Fornitore: Apollo Scientific

PC303372-1G PC303372-5G
APOSPC303372-1GEA 339 EUR
APOSPC303372-1G APOSPC303372-5G
4,6-Dichloro-2-fluoro-3-methoxybenzylamine
4,6-Dichloro-2-fluoro-3-methoxybenzylamine
Formula: C₈H₈Cl₂FNO
Peso molecolare: 224,06 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD30726940
CAS Number: 2149590-99-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR