Stampa…

2,4-Difluoro-5-methylcinnamic acid 97%

Fornitore: Apollo Scientific

PC303312-5G PC303312-1G
APOSPC303312-5GEA 902 EUR
APOSPC303312-5G APOSPC303312-1G
2,4-Difluoro-5-methylcinnamic acid 97%
2,4-Difluoro-5-methylcinnamic acid
Formula: C₁₀H₈F₂O₂
Peso molecolare: 198,166 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD23757123
CAS Number: 1562311-33-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR