Stampa…

2,4-Difluoro-5-methylbenzyl bromide 97%

Fornitore: Apollo Scientific
Pericolo

PC303303-1G PC303303-5G
APOSPC303303-1GEA 306 EUR
APOSPC303303-1G APOSPC303303-5G
2,4-Difluoro-5-methylbenzyl bromide 97%
2,4-Difluoro-5-methylbenzyl bromide
Formula: C₈H₇BrF₂
Peso molecolare: 221,042 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD19440200
CAS Number: 315204-37-6
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR