Stampa…

3,4-Dichloro-5-(trifluoromethyl)benzamide 97%

Fornitore: Apollo Scientific

PC303176-1G PC303176-250MG
APOSPC303176-1GEA 674 EUR
APOSPC303176-1G APOSPC303176-250MG
3,4-Dichloro-5-(trifluoromethyl)benzamide 97%
3,4-Dichloro-5-(trifluoromethyl)benzamide
Formula: C₈H₄Cl₂F₃NO
Peso molecolare: 258,025 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28054200
CAS Number: 1706446-52-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR