2,4,5-Trifluoro-DL-phenylalanine
Fornitore: Apollo Scientific
PC302433-1G
PC302433-5G
PC302433-10G
APOSPC302433-1GEA
118
EUR
APOSPC302433-1G
APOSPC302433-5G
APOSPC302433-10G
2,4,5-Trifluoro-DL-phenylalanine
2,4,5-Trifluoro-DL-phenylalanine
Formula:
C₉H₈F₃NO₂ Peso molecolare: 219,161 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04116039 CAS Number: 1260002-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...