Stampa…

4-Chloro-3-(trifluoromethoxy)cinnamic acid 97%

Fornitore: Apollo Scientific

PC302059-1G PC302059-5G
APOSPC302059-1GEA 105 EUR
APOSPC302059-1G APOSPC302059-5G
4-Chloro-3-(trifluoromethoxy)cinnamic acid 97%
4-Chloro-3-(trifluoromethoxy)cinnamic acid
Formula: C₁₀H₆ClF₃O₃
Peso molecolare: 266,601 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06660262
CAS Number: 886501-56-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR