4-Chloro-4,4-difluoro-1-phenylbutane-1,3-dione
Fornitore: Apollo Scientific
|
Attenzione
|
PC28292-1G
PC28292-250MG
PC28292-5G
APOSPC28292-1GEA
285
EUR
APOSPC28292-1G
APOSPC28292-250MG
APOSPC28292-5G
4-Chloro-4,4-difluoro-1-phenylbutane-1,3-dione
4-Chloro-4,4-difluoro-1-phenylbutane-1,3-dione
Formula:
C₁₀H₇ClF₂O₂ Peso molecolare: 232,611 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03420658 CAS Number: 2062-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...