Stampa…

2-Bromo-6-nitrobenzyl fluoride 95%

Fornitore: Apollo Scientific
Pericolo

PC27901-1G PC27901-250MG
APOSPC27901-1GEA 611 EUR
APOSPC27901-1G APOSPC27901-250MG
2-Bromo-6-nitrobenzyl fluoride 95%
2-Bromo-6-nitrobenzyl fluoride
Formula: C₇H₅BrFNO₂
Peso molecolare: 234,023 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD31664894
CAS Number: 2244088-13-7
UN: 3261
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR