Stampa…

3-Cyano-2,6-difluorobenzoic acid

Fornitore: Apollo Scientific

PC210042-1G PC210042-500MG
APOSPC210042-1GEA 727 EUR
APOSPC210042-1G APOSPC210042-500MG
3-Cyano-2,6-difluorobenzoic acid
3-Cyano-2,6-difluorobenzoic acid
Formula: C₈H₃F₂NO₂
Peso molecolare: 183,112 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09907851
CAS Number: 1314971-48-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR