1-Fluoro-3-(2-nitrovinyl)benzene 95+%
Fornitore: Apollo Scientific
PC201286-1G
PC201286-25G
PC201286-5G
APOSPC201286-1GEA
68.4
EUR
APOSPC201286-1G
APOSPC201286-25G
APOSPC201286-5G
1-Fluoro-3-(2-nitrovinyl)benzene 95+%
1-Fluoro-3-(2-nitrovinyl)benzene
Formula:
C₈H₆FNO₂ Peso molecolare: 167,137 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00176732 CAS Number: 705-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...