5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole 96%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
2-Bromo-4,5-(difluoromethylenedioxy)aniline, 6-Bromo-2,2-difluorobenzo[d][1,3]dioxol-5-amine, 6-Bromo-2,2-difluoro-1,3-benzodioxol-5-amine
PC1978-1G
PC1978-5G
APOSPC1978-1GEA
94.5
EUR
APOSPC1978-1G
APOSPC1978-5G
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole 96%
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole
Formula:
C₇H₄BrF₂NO₂ Peso molecolare: 252,013 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04972651 CAS Number: 887267-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...