tert-Butyl trifluoroacetate 99%
Fornitore: Apollo Scientific
|
Attenzione
|
Synonyms:
tert-Butyl 2,2,2-trifluoroethanoate, Trifluoroacetic acid tert-butyl ester
PC0694-5G
PC0694-25G
APOSPC0694-5GEA
87.3
EUR
APOSPC0694-5G
APOSPC0694-25G
tert-Butyl trifluoroacetate 99%
tert-Butyl trifluoroacetate
Formula:
C₆H₉F₃O₂ Peso molecolare: 170,13 g/mol Punto di ebollizione: 83 °C Densità: 1,089 g/cm³ (25 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00780927 CAS Number: 400-52-2 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...