Ethyl 3-(2-aminophenyl)acrylate 98%
Fornitore: Apollo Scientific
OR962902-1G
OR962902-5G
APOSOR962902-1GEA
248
EUR
APOSOR962902-1G
APOSOR962902-5G
Ethyl 3-(2-aminophenyl)acrylate 98%
Ethyl 3-(2-aminophenyl)acrylate
Formula:
C₁₁H₁₃NO₂ Peso molecolare: 191,226 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD12407284 CAS Number: 79655-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...