Stampa…

tert-Butyl 3-bromophenethylcarbamate 98%

Fornitore: Apollo Scientific
Attenzione

OR962835-1G OR962835-5G
APOSOR962835-1GEA 198 EUR
APOSOR962835-1G APOSOR962835-5G
tert-Butyl 3-bromophenethylcarbamate 98%
tert-Butyl 3-bromophenethylcarbamate
Formula: C₁₃H₁₈BrNO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11042237
CAS Number: 153732-25-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR