Stampa…

(2-Methyl-quinolin-4-yl)-methanol 95%

Fornitore: Apollo Scientific

Synonyms: (2-Methylquinolin-4-yl)methanol

OR962675-1G OR962675-250MG OR962675-5G
APOSOR962675-1GEA 234 EUR
APOSOR962675-1G APOSOR962675-250MG APOSOR962675-5G
(2-Methyl-quinolin-4-yl)-methanol 95%
(2-Methyl-quinolin-4-yl)-methanol
Formula: C₁₁H₁₁NO
Peso molecolare: 173,211 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD07371472
CAS Number: 4939-28-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR