Stampa…

Ethyl 6-methylpyridine-3-carboxylate 95%

Fornitore: Apollo Scientific

Synonyms: Ethyl 6-methylnicotinate

OR962497-1G OR962497-5G
APOSOR962497-1GEA 59.7 EUR
APOSOR962497-1G APOSOR962497-5G
Ethyl 6-methylpyridine-3-carboxylate 95%
Ethyl 6-methylpyridine-3-carboxylate
Formula: C₉H₁₁NO₂
Peso molecolare: 165,189 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00234551
CAS Number: 21684-59-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR