Stampa…

3-(Boc-amino)-2-phenylpropanoic acid 97%

Fornitore: Apollo Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-2-phenylpropanoic acid

OR961404-1G OR961404-500MG
APOSOR961404-1GEA 426 EUR
APOSOR961404-1G APOSOR961404-500MG
3-(Boc-amino)-2-phenylpropanoic acid 97%
3-(Boc-amino)-2-phenylpropanoic acid
Formula: C₁₄H₁₉NO₄
Peso molecolare: 265,305 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03003692
CAS Number: 67098-56-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR