2-amino-4-isopropylthiophene-3-carbonitrile 96%
Fornitore: Apollo Scientific
Formula:
C₈H₁₀N₂S Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD10695775 CAS Number: 10413-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...