4-(BOC-Amino)-1-isopentylpiperidine 98%
Fornitore: Apollo Scientific
|
Attenzione
|
Formula:
C₁₅H₃₀N₂O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD21333075 CAS Number: 888944-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...