N-Methoxy-N-methylhexanamide 96%
Fornitore: Apollo Scientific
Formula:
C₈H₁₇NO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD14707560 CAS Number: 64214-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...