6-Cyclohexyl-4-methyl-2H-pyran-2-one 95%
Fornitore: Apollo Scientific
OR958317-100G
OR958317-25G
APOSOR958317-100GEA
379
EUR
APOSOR958317-100G
APOSOR958317-25G
6-Cyclohexyl-4-methyl-2H-pyran-2-one 95%
6-Cyclohexyl-4-methyl-2H-pyran-2-one
Formula:
C₁₂H₁₆O₂ Peso molecolare: 192,254 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD09953433 CAS Number: 14818-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...