3-Hydroxy-5-methylbenzaldehyde 96%
Fornitore: Apollo Scientific
OR957500-1G
OR957500-5G
APOSOR957500-1GEA
183
EUR
APOSOR957500-1G
APOSOR957500-5G
3-Hydroxy-5-methylbenzaldehyde 96%
3-Hydroxy-5-methylbenzaldehyde
Formula:
C₈H₈O₂ Peso molecolare: 136,148 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD16999707 CAS Number: 60549-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...