Stampa…

3-Acetamido-4-methylbenzoic acid 98%

Fornitore: Apollo Scientific

OR954620-10G OR954620-25G OR954620-100G
APOSOR954620-10GEA 190 EUR
APOSOR954620-10G APOSOR954620-25G APOSOR954620-100G
3-Acetamido-4-methylbenzoic acid 98%
3-Acetamido-4-methylbenzoic acid
Formula: C₁₀H₁₁NO₃
Peso molecolare: 193,199 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00020381
CAS Number: 6946-14-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR