Stampa…

1,2-Dimethylbenzodiazole-5-carboxylic acid 98%

Fornitore: Apollo Scientific

OR954338-1G OR954338-5G
APOSOR954338-1GEA 219 EUR
APOSOR954338-1G APOSOR954338-5G
1,2-Dimethylbenzodiazole-5-carboxylic acid 98%
1,2-Dimethylbenzodiazole-5-carboxylic acid
Formula: C₁₀H₁₀N₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01240945
CAS Number: 90915-18-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR