4-Phenoxybutan-1-amine 96%
Fornitore: Apollo Scientific
Formula:
C₁₀H₁₅NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00031400 CAS Number: 16728-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...