Stampa…

9-Oxo-bicyclo[3.3.1]nonane-3-carboxylic acid 95%

Fornitore: Apollo Scientific

OR953962-1G OR953962-250MG
APOSOR953962-1GEA 364 EUR
APOSOR953962-1G APOSOR953962-250MG
9-Oxo-bicyclo[3.3.1]nonane-3-carboxylic acid 95%
9-Oxo-bicyclo[3.3.1]nonane-3-carboxylic acid
Formula: C₁₀H₁₄O₃
Peso molecolare: 182,216 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD08447209
CAS Number: 63242-00-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR