Stampa…

Ethyl 5-methylpyrazine-2-carboxylate 95%

Fornitore: Apollo Scientific

OR952290-10G OR952290-1G OR952290-25G
APOSOR952290-10GEA 101 EUR
APOSOR952290-10G APOSOR952290-1G APOSOR952290-25G
Ethyl 5-methylpyrazine-2-carboxylate 95%
Ethyl 5-methylpyrazine-2-carboxylate
Formula: C₈H₁₀N₂O₂
Peso molecolare: 166,177 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD05662506
CAS Number: 41110-34-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR