Stampa…

4-Amino-2,5-dichloropyrimidine 95%

Fornitore: Apollo Scientific

Synonyms: 2,5-Dichloropyrimidin-4-amine

OR951944-10G OR951944-1G OR951944-5G
APOSOR951944-10GEA 328 EUR
APOSOR951944-10G APOSOR951944-1G APOSOR951944-5G
4-Amino-2,5-dichloropyrimidine 95%
4-Amino-2,5-dichloropyrimidine
Formula: C₄H₃Cl₂N₃
Peso molecolare: 163,993 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD13191633
CAS Number: 89180-51-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR