Stampa…

Methyl 1,2,3-triazole-4-carboxylate 95%

Fornitore: Apollo Scientific
Attenzione

Synonyms: Methyl 1H-1,2,3-triazole-4-carboxylate

OR951706-1G OR951706-250MG
APOSOR951706-1GEA 75.7 EUR
APOSOR951706-1G APOSOR951706-250MG
Methyl 1,2,3-triazole-4-carboxylate 95%
Methyl 1,2,3-triazole-4-carboxylate
Formula: C₄H₅N₃O₂
Peso molecolare: 127,101 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD12912989
CAS Number: 4967-77-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR