Stampa…
Cycloheptylmethanamine hydrochloride 95%
Fornitore: Apollo Scientific
OR950869-10G OR950869-1G OR950869-5G
APOSOR950869-10GEA 275 EUR
APOSOR950869-10G APOSOR950869-1G APOSOR950869-5G
Cycloheptylmethanamine hydrochloride 95%
Cycloheptylmethanamine hydrochloride
Formula: C₈H₁₇N·ClH
Peso molecolare: 163.688 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD23701552
CAS Number: 177352-26-0