Stampa…

2-(Hydroxymethyl)benzoic acid 95%

Fornitore: Apollo Scientific

OR947220-1G OR947220-250MG
APOSOR947220-1GEA 865 EUR
APOSOR947220-1G APOSOR947220-250MG
2-(Hydroxymethyl)benzoic acid 95%
2-(Hydroxymethyl)benzoic acid
Formula: C₈H₈O₃
Peso molecolare: 152,147 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD02151998
CAS Number: 612-20-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR