2-Methyl-2-morpholinopropanenitrile 95%
Fornitore: Apollo Scientific
Formula:
C₈H₁₄N₂O |
MDL Number:
MFCD04639674 CAS Number: 35666-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...