Stampa…

Tropine-3-thiol hydrochloride 95%

Fornitore: Apollo Scientific

OR944399-1G OR944399-250MG
APOSOR944399-1GEA 771 EUR
APOSOR944399-1G APOSOR944399-250MG
Tropine-3-thiol hydrochloride 95%
Tropine-3-thiol hydrochloride
Formula: C₈H₁₅NS·ClH
Peso molecolare: 193,737 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12965024
CAS Number: 908266-48-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR