3,4-Dihydro-1,4-benzoxazepin-5(2H)-one 95%
Fornitore: Apollo Scientific
Formula:
C₉H₉NO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02726416 CAS Number: 703-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...