Stampa…

(R)-1-(3-Nitrophenyl)ethanamine 95%

Fornitore: Apollo Scientific

OR943497-1G OR943497-250MG
APOSOR943497-1GEA 828 EUR
APOSOR943497-1G APOSOR943497-250MG
(R)-1-(3-Nitrophenyl)ethanamine 95%
(R)-1-(3-Nitrophenyl)ethanamine
Formula: C₈H₁₀N₂O₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04972379
CAS Number: 297730-27-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR