Stampa…

4-Bromo-2-methylbutan-2-ol 95%

Fornitore: Apollo Scientific

Synonyms: 4-Bromo-2-methyl-2-butanol

OR939949-1G OR939949-250MG
APOSOR939949-1GEA 141 EUR
APOSOR939949-1G APOSOR939949-250MG
4-Bromo-2-methylbutan-2-ol 95%
4-Bromo-2-methylbutan-2-ol
Formula: C₅H₁₁BrO
Peso molecolare: 167,044 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD20483824
CAS Number: 35979-69-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR