Stampa…

O-(3-Chloroallyl)hydroxylamine 95%

Fornitore: Apollo Scientific

Synonyms: (E)-O-(3-Chloroallyl)hydroxylamine

OR939572-10G OR939572-25G
APOSOR939572-10GEA 212 EUR
APOSOR939572-10G APOSOR939572-25G
O-(3-Chloroallyl)hydroxylamine 95%
O-(3-Chloroallyl)hydroxylamine
Formula: C₃H₆ClNO
Peso molecolare: 107,539 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09951937
CAS Number: 87851-77-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR