Stampa…

1-Acetyl-1-cyclopentene 95%

Fornitore: Apollo Scientific

Synonyms: 1-(Cyclopent-1-en-1-yl)ethanone

OR939571-1G OR939571-5G
APOSOR939571-1GEA 285 EUR
APOSOR939571-1G APOSOR939571-5G
1-Acetyl-1-cyclopentene 95%
1-Acetyl-1-cyclopentene
Formula: C₇H₁₀O
Peso molecolare: 110,154 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00799464
CAS Number: 16112-10-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR