Stampa…

Ethyl 5-methylnicotinate 95%

Fornitore: Apollo Scientific

OR939551-1G OR939551-5G
APOSOR939551-1GEA 53.9 EUR
APOSOR939551-1G APOSOR939551-5G
Ethyl 5-methylnicotinate 95%
Ethyl 5-methylnicotinate
Formula: C₉H₁₁NO₂
Peso molecolare: 165,189 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09030156
CAS Number: 20826-02-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR